• USA Home
  • Search Results

  • brilliantbluerconcentrate82597610459211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Brilliant Blue R Concentrate

1 Products
Synonym:   Acid Blue 83, Brilliant Blue R, Brilliant indocyanin 6B, Coomassie Brilliant Blue R
Image of Brilliant Blue R Concentrate
CAS Number:
Empirical Formula (Hill Notation):
 C45H44N3NaO7S2
Molecular Weight:
 825.97
 Beilstein/REAXYS Number:
 5718025
 Colour Index Number:
 42660
 MDL number:
 NACRES:
 NA.47
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More