• USA Home
  • Search Results

  • bromine15981772695611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Bromine

7 Products
Image of Bromine
CAS Number:
Empirical Formula (Hill Notation):
 Br2
Molecular Weight:
 159.81
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Restriction
  Viscosity
  Density
  Vapor pressure
  Solubility
  Melting Point
  Assay
Show More