• USA Home
  • Search Results

  • bromochloroaceticacid17339558996811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Bromochloroacetic acid

1 Products
Image of Bromochloroacetic acid
CAS Number:
Linear Formula:
 BrClCHCOOH
Molecular Weight:
 173.39
 Beilstein/REAXYS Number:
 1720556
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More