• USA Home
  • Search Results

  • bromocresolpurple5402211540211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Bromocresol purple

4 Products
Synonym:   5′,5′′-Dibromo-o-cresolsulfonphthalein, Bromocresol purple
Image of Bromocresol purple
CAS Number:
Empirical Formula (Hill Notation):
 C21H16Br2O5S
Molecular Weight:
 540.22
 EC Index Number:
 204-087-8
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Absorption
  Product Line
  Solubility
  Physical Form
  Special Grade
Show More