• USA Home
  • Search Results

  • bromotrimethylsilane15309285797811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Bromotrimethylsilane

3 Products
Synonym:   TMBS, Trimethylbromosilane, Trimethylsilyl bromide
Image of Bromotrimethylsilane
CAS Number:
Linear Formula:
 (CH3)3SiBr
Molecular Weight:
 153.09
 Beilstein/REAXYS Number:
 1731135
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Additive
Show More