• USA Home
  • Search Results

  • bronopol199995251711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Bronopol

1 Products
Synonym:   2-Bromo-2-nitro-1,3-propanediol, BNPD, BNPK, Bronopol
Image of Bronopol
CAS Number:
Linear Formula:
 HOCH2C(Br)(NO2)CH2OH
Molecular Weight:
 199.99
 Beilstein/REAXYS Number:
 1705868
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More