• USA Home
  • Search Results

  • brucine3944635757311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Brucine

3 Products
Synonym:   10,11-Dimethoxystrychnine, 2,3-Dimethoxystrychnidin-10-one
Image of Brucine
CAS Number:
Empirical Formula (Hill Notation):
 C23H26N2O4
Molecular Weight:
 394.46
 Beilstein/REAXYS Number:
 63046
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Manufacturer designation
  Assay
Show More