• USA Home
  • Search Results

  • brukerstandardsolvent114027605111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Bruker standard solvent

1 Products
Synonym:   TFA in H2O/ACN, TFA, Trifluoroacetic acid solution
Image of Bruker standard solvent
CAS Number:
Linear Formula:
 CF3COOH
Molecular Weight:
 114.02
 Beilstein/REAXYS Number:
 5446934
 MDL number:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Additive
Show More