• USA Home
  • Search Results

  • bsitosterol414718346511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

β-Sitosterol

9 Products
Synonym:   α-Dihydrofucosterol, 22,23-Dihydrostigmasterol, 24α-Ethylcholesterol, 5-Stigmasten-3β-ol, beta-Sitosterol
Image of β-Sitosterol
CAS Number:
Empirical Formula (Hill Notation):
 C29H50O
Molecular Weight:
 414.71
 Beilstein/REAXYS Number:
 1916165
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More