• USA Home
  • Search Results

  • bthujaplicin1642049944511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

β-Thujaplicin

1 Products
Synonym:   2-Hydroxy-4-isopropyl-2,4,6-cycloheptatrien-1-one, Hinokitiol, beta-Thujaplicin
Image of β-Thujaplicin
CAS Number:
Empirical Formula (Hill Notation):
 C10H12O2
Molecular Weight:
 164.20
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
469521 β-Thujaplicin, 99%,