• USA Home
  • Search Results

  • bulleyaconitinea6277610766879111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Bulleyaconitine A

1 Products
Synonym:   ((1-α,6-α,14-α,16-β)-8-(acetyloxy)-20-ethyl-13-hydroxy-1,6,16-trimethoxy-4-(methoxymethyl)aconitan-14-yl)(4-methoxyphenyl)-methanone
Image of Bulleyaconitine A
CAS Number:
Empirical Formula (Hill Notation):
 C35H49NO9
Molecular Weight:
 627.76
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More