• USA Home
  • Search Results

  • bupropionhydrochloriderelatedcompoundb32065104971843511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Bupropion Hydrochloride Related Compound B

2 Products
Synonym:   2-(tert-Butylamino)-3′-bromopropiophenone hydrochloride, 2-(tert-butylamino)-3’-bromopiophenone hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C13H18BrNO · HCl
Molecular Weight:
 320.65
 NACRES
 NA.24
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Application(s)
Show More