• USA Home
  • Search Results

  • buspironehydrochloride421963338608211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Buspirone hydrochloride

4 Products
Synonym:   N-[4-[4-(2-Pyrimidinyl)-1-piperazinyl]butyl]-8-azaspiro[4.5]decane-7,9-dione hydrochloride
Image of Buspirone hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C21H31N5O2 · HCl
Molecular Weight:
 421.96
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Product Information Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Manufacturer designation
Show More