• USA Home
  • Search Results

  • butalbitalsolution224267726911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Butalbital solution

1 Products
Synonym:   5-Allyl-5-isobutylbarbituric acid
Image of Butalbital solution
CAS Number:
Empirical Formula (Hill Notation):
 C11H16N2O3
Molecular Weight:
 224.26
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More