• USA Home
  • Search Results

  • butane581210697811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Butane

1 Products
Synonym:   n-Butane
Image of Butane
CAS Number:
Linear Formula:
 CH3CH2CH2CH3
Molecular Weight:
 58.12
 Beilstein/REAXYS Number:
 969129
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
494402 Butane, 99%,