• USA Home
  • Search Results

  • butyl2methylbutyrate158241570673711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Butyl 2-methylbutyrate

4 Products
Image of Butyl 2-methylbutyrate
CAS Number:
Linear Formula:
 CH3CH2CH(CH3)COOCH2CH2CH2CH3
Molecular Weight:
 158.24
 Beilstein Registry Number:
 1702551
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Document Links
  Allergen
  Organoleptic property
  Titertest
  Refractive Index
  Special Grade
  Assay
Show More