• USA Home
  • Search Results

  • butylacrylate1281714132211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Butyl acrylate

3 Products
Image of Butyl acrylate
CAS Number:
Linear Formula:
 CH2=CHCOO(CH2)3CH3
Molecular Weight:
 128.17
 Beilstein/REAXYS Number:
 1749970
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Refractive Index
  Special Grade
  Additive
  Density
Show More