• USA Home
  • Search Results

  • butylalcohol74127136311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Butyl alcohol

2 Products
Synonym:   1-Butanol, n-Butanol, Butyl alcohol
Image of Butyl alcohol
CAS Number:
Linear Formula:
 CH3(CH2)3OH
Molecular Weight:
 74.12
 Beilstein/REAXYS Number:
 969148
 Council of Europe no.:
 52c
 EC Number:
 FEMA Number:
 2178
 Flavis number:
 2.004
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Biological Source
  Document Links
  Allergen
  Organoleptic property
  Assay
  Special Grade
Show More