• USA Home
  • Search Results

  • butylatedhydroxyanisole180242501316511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Butylated hydroxyanisole

5 Products
Synonym:   2(3)-t-Butyl-4-hydroxyanisole, 2(3)-t-Butylhydroquinone monomethyl ether, BHA
Image of Butylated hydroxyanisole
CAS Number:
Linear Formula:
 (CH3)3CC6H3(OCH3)OH
Molecular Weight:
 180.24
 EC Number:
 MDL number:
 NACRES:
 NA.22
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Shelf Life
  Physical Form
  Assay
  Agency/Method
  Application(s)
Show More