• USA Home
  • Search Results

  • butylatedhydroxytoluene2203512837011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Butylated hydroxytoluene

3 Products
Synonym:   2,6-Di-tert-butyl-4-methylphenol, 2,6-Di-tert-butyl-p-cresol, BHT, Butylated hydroxytoluene, Butylhydroxytoluene, DBPC
Image of Butylated hydroxytoluene
CAS Number:
Linear Formula:
 [(CH3)3C]2C6H2(CH3)OH
Molecular Weight:
 220.35
 Beilstein/REAXYS Number:
 1911640
 E Number:
 E 321
 EC Number:
 FEMA Number:
 2184
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Application(s)
  Physical Form
  Agency/Method
Show More