• USA Home
  • Search Results

  • butylisothiocyanate1152059282511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Butyl isothiocyanate

3 Products
Synonym:   NSC 194808
Image of Butyl isothiocyanate
CAS Number:
Linear Formula:
 CH3(CH2)3NCS
Molecular Weight:
 115.20
 Beilstein/REAXYS Number:
 906839
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
Show More