• USA Home
  • Search Results

  • butyllaurate2564210618311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Butyl laurate

1 Products
Synonym:   Butyl dodecanoate, Butyl laurate, Lauric acid butyl ester
Image of Butyl laurate
CAS Number:
Linear Formula:
 CH3(CH2)10COO(CH2)3CH3
Molecular Weight:
 256.42
 Beilstein/REAXYS Number:
 1776360
 Council of Europe no.:
 376
 EC Number:
 FEMA Number:
 2206
 Flavis number:
 9.100
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
W220604 Butyl laurate, ≥99%,