• USA Home
  • Search Results

  • butylmagnesiumchloridesolution1168769304911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Butylmagnesium chloride solution

3 Products
Synonym:   BuMgCl
Image of Butylmagnesium chloride solution
CAS Number:
Linear Formula:
 CH3(CH2)3MgCl
Molecular Weight:
 116.87
 Beilstein/REAXYS Number:
 3587228
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Density
Show More