• USA Home
  • Search Results

  • butylstearate3405812395511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Butyl stearate

3 Products
Synonym:   Stearic acid butyl ester
Image of Butyl stearate
CAS Number:
Linear Formula:
 CH3(CH2)16COO(CH2)3CH3
Molecular Weight:
 340.58
 Beilstein/REAXYS Number:
 1792866
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
Show More