• USA Home
  • Search Results

  • butylsulfide1462954440111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Butyl sulfide

1 Products
Synonym:   1,1′-Thiobisbutane, Butyl sulfide, Dibutyl sulfide, NSC 8460
Image of Butyl sulfide
CAS Number:
Linear Formula:
 CH3(CH2)3S(CH2)3CH3
Molecular Weight:
 146.29
 Beilstein Registry Number:
 1732829
 Council of Europe no.:
 484
 EC Number:
 FEMA Number:
 2215
 Flavis number:
 12.007
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
Show More