• USA Home
  • Search Results

  • butyricacid881110792611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Butyric acid

5 Products
Image of Butyric acid
CAS Number:
Empirical Formula (Hill Notation):
 C4H8O2
Molecular Weight:
 88.11
 EC Index Number
 607-135-00-X
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Agency/Method
  Document Links
  Allergen
  Organoleptic property
  Titertest
  Assay
  Special Grade
Show More