• USA Home
  • Search Results

  • cacodylicacid138007560511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cacodylic acid

2 Products
Synonym:   Dimethylarsinic acid, Dimethylarsonic acid, Hydroxydimethylarsine oxide
Image of Cacodylic acid
CAS Number:
Linear Formula:
 (CH3)2As(O)OH
Molecular Weight:
 138.00
 Beilstein/REAXYS Number:
 1736965
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Titertest
Show More