• USA Home
  • Search Results

  • cadmium11241744043911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cadmium

223 Products
Image of Cadmium
CAS Number:
Empirical Formula (Hill Notation):
 Cd
Molecular Weight:
 112.41
 Beilstein/REAXYS Number:
 8137359
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Vapor pressure
  Melting Point
  Density
  Reaction Suitability
  Weight
  Particle Dimension
  Composite Dimension
  Manufacturer designation
  Physical Form
Show More

Product #
Description
265365 Cadmium, powder, −100 mesh, 99.5% trace metals basis,
265330 Cadmium, granular, ≥99%, 5-20 mesh,
414891 Cadmium, granular, 30-80 mesh, ≥99%,
1.02001 Cadmium, coarse powder, for analysis and for filling reductors particle size about 0.3-1.6 mm EMSURE®,
265454 Cadmium, rod, diam. 4.0 mm, 99.999% trace metals basis,
202886 Cadmium, shot, 3 mm, 99.999% trace metals basis,
GF58541272 Cadmium, powder, max. particle size 60 micron, weight 100 g, purity 99.9%,
1.02004 Cadmium, granular, for analysis particle size about 3-6 mm EMSURE®,
GF61633188 Cadmium, lump, 5 mm max. lump size, weight 50 g, purity 99.99+%,
GF70277766 Cadmium, lump, 100 mm max. lump size, weight 1000 g, purity 99.99%,
GF98162220 Cadmium, lump, 100 mm max. lump size, weight 500 g, purity 99.99%,
GF04148265 Cadmium, lump, 5 mm max. lump size, weight 100 g, purity 99.99+%,
GF07484575 Cadmium, powder, max. particle size 60 micron, weight 500 g, purity 99.9%,
GF16785298 Cadmium, lump, 100 mm max. lump size, weight 200 g, purity 99.99%,
GF17659873 Cadmium, powder, max. particle size 60 micron, weight 200 g, purity 99.9%,
GF18533527 Cadmium, lump, 5 mm max. lump size, weight 200 g, purity 99.99+%,
GF35572271 Cadmium, powder, max. particle size 60 micron, weight 1000 g, purity 99.9%,
GF46534282 Cadmium, lump, 100 mm max. lump size, weight 100 g, purity 99.99%,
GF53886358 Cadmium, wire reel, 0.1m, diameter 0.70mm, as drawn, 99.99+%,
GF82400857 Cadmium, wire reel, 0.1m, diameter 1.2mm, as drawn, 99.99+%,
GF24318794 Cadmium, wire reel, 0.1m, diameter 2.0mm, hard, 99.99+%,
GF42263849 Cadmium, wire reel, 0.1m, diameter 1.0mm, hard, 99.999%,
GF12224078 Cadmium, wire reel, 0.2m, diameter 0.25mm, hard, 99.99+%,
GF23019960 Cadmium, wire reel, 0.2m, diameter 0.5mm, hard, 99.95+%,
GF37213893 Cadmium, wire reel, 0.2m, diameter 0.5mm, hard, 99.99+%,
GF37063144 Cadmium, wire reel, 0.2m, diameter 0.70mm, as drawn, 99.99+%,
GF80208536 Cadmium, wire reel, 0.2m, diameter 1.2mm, as drawn, 99.99+%,
GF39849935 Cadmium, wire reel, 0.2m, diameter 2.0mm, hard, 99.99+%,
GF38869105 Cadmium, wire reel, 0.2m, diameter 0.125mm, as drawn, 99.9%,
GF82981064 Cadmium, wire reel, 0.2m, diameter 1.0mm, hard, 99.999%,
GF28009593 Cadmium, wire reel, 0.5m, diameter 0.25mm, hard, 99.99+%,
GF77495833 Cadmium, wire reel, 0.5m, diameter 0.5mm, hard, 99.95+%,
GF51736653 Cadmium, wire reel, 0.5m, diameter 0.5mm, hard, 99.99+%,
GF12560868 Cadmium, wire reel, 0.5m, diameter 0.70mm, as drawn, 99.99+%,
GF55395298 Cadmium, wire reel, 0.5m, diameter 1.2mm, as drawn, 99.99+%,
GF81241523 Cadmium, wire reel, 0.5m, diameter 2.0mm, hard, 99.99+%,
GF58637762 Cadmium, wire reel, 0.5m, diameter 0.125mm, as drawn, 99.9%,
GF72632998 Cadmium, wire reel, 0.5m, diameter 1.0mm, hard, 99.999%,
GF30757383 Cadmium, wire reel, 10m, diameter 0.25mm, hard, 99.99+%,
GF01198762 Cadmium, wire reel, 1m, diameter 0.25mm, hard, 99.99+%,
GF09930359 Cadmium, foil, 100x100mm, thickness 0.125mm, as rolled, 99.95%,
GF50870482 Cadmium, foil, 100x100mm, thickness 0.125mm, as rolled, 99.99+%,
GF61936814 Cadmium, foil, 100x100mm, thickness 0.25mm, as rolled, 99.99+%,
GF64286602 Cadmium, foil, 100x100mm, thickness 1.0mm, as rolled, 99.99%,
GF30096345 Cadmium, foil, 100x100mm, thickness 2.0mm, as rolled, 99.99%,
GF10741889 Cadmium, foil, 100x100mm, thickness 3.0mm, as rolled, 99.9%,
GF37649100 Cadmium, foil, 100x200mm, thickness 0.125mm, as rolled, 99.95%,
GF38583287 Cadmium, foil, 100x300mm, thickness 0.125mm, as rolled, 99.95%,
GF50990597 Cadmium, foil, 150x150mm, thickness 0.25mm, as rolled, 99.95%,
GF72757503 Cadmium, foil, 150x150mm, thickness 0.5mm, as rolled, 99.99%,
GF66037754 Cadmium, foil, 150x150mm, thickness 1.0mm, as rolled, 99.95%,
GF76768914 Cadmium, foil, 150x150mm, thickness 3.0mm, as rolled, 99.9%,
GF33512688 Cadmium, foil, 200x200mm, thickness 0.5mm, as rolled, 99.99%,
GF94010453 Cadmium, foil, 200x200mm, thickness 1.0mm, as rolled, 99.99%,
GF90633652 Cadmium, foil, 25x25mm, thickness 0.125mm, as rolled, 99.99+%,
GF06836238 Cadmium, foil, 25x25mm, thickness 0.25mm, as rolled, 99.9%,
GF71408763 Cadmium, foil, 25x25mm, thickness 0.25mm, as rolled, 99.99+%,
GF59435062 Cadmium, foil, 25x25mm, thickness 0.5mm, as rolled, 99.999%,
GF78263411 Cadmium, foil, 25x25mm, thickness 1.0mm, as rolled, 99.999%,
GF84554613 Cadmium, foil, 300x1000mm, thickness 2.0mm, as rolled, 99.99%,
GF19008817 Cadmium, foil, 300x300mm, thickness 0.25mm, as rolled, 99.95%,
GF20958947 Cadmium, foil, 300x300mm, thickness 0.5mm, as rolled, 99.99%,
GF15879954 Cadmium, foil, 300x300mm, thickness 2.0mm, as rolled, 99.99%,
GF72851182 Cadmium, foil, 30x100mm, thickness 0.5mm, as rolled, 99.999%,
GF06398039 Cadmium, foil, 30x150mm, thickness 0.5mm, as rolled, 99.999%,
GF29382802 Cadmium, foil, 30x50mm, thickness 0.5mm, as rolled, 99.999%,
GF65882773 Cadmium, foil, 50x100mm, thickness 1.0mm, as rolled, 99.999%,
GF35346281 Cadmium, foil, 50x200mm, thickness 1.0mm, as rolled, 99.999%,
GF07279443 Cadmium, foil, 50x50mm, thickness 0.125mm, as rolled, 99.95%,
GF29206964 Cadmium, foil, 50x50mm, thickness 0.125mm, as rolled, 99.99+%,
GF54529331 Cadmium, foil, 50x50mm, thickness 0.25mm, as rolled, 99.9%,
GF26095495 Cadmium, foil, 50x50mm, thickness 0.25mm, as rolled, 99.99+%,
GF68941390 Cadmium, foil, 50x50mm, thickness 1.0mm, as rolled, 99.95%,
GF82927064 Cadmium, foil, 50x50mm, thickness 1.0mm, as rolled, 99.999%,
GF52240777 Cadmium, foil, 50x50mm, thickness 2.0mm, as rolled, 99.99%,
GF36085934 Cadmium, foil, 50x50mm, thickness 3.0mm, as rolled, 99.9%,
GF97050652 Cadmium, foil, 75x100mm, thickness 0.5mm, as rolled, 99.99%,
GF13392738 Cadmium, foil, 95x100mm, thickness 0.5mm, as rolled, 99.99%,
GF99754161 Cadmium, foil, 0.5m coil, thickness 0.5mm, as rolled, 100%,
GF52168608 Cadmium, foil, 10mm disks, thickness 0.005mm, 99.7+%,
GF01095195 Cadmium, foil, 10mm disks, thickness 0.015mm, 99.7+%,
GF82714091 Cadmium, foil, 10mm disks, thickness 0.01mm, 99.7+%,
GF89972791 Cadmium, foil, 10mm disks, thickness 0.025mm, 99.7+%,
GF63705887 Cadmium, foil, 10mm disks, thickness 0.05mm, as rolled, 99.99+%,
GF99067595 Cadmium, foil, 10mm disks, thickness 0.125mm, as rolled, 99.95%,
GF60235219 Cadmium, foil, 10mm disks, thickness 0.125mm, as rolled, 99.99+%,
GF73871107 Cadmium, foil, 10mm disks, thickness 0.25mm, as rolled, 99.9%,
GF93011396 Cadmium, foil, 10mm disks, thickness 0.25mm, as rolled, 99.95%,
GF33674777 Cadmium, foil, 10mm disks, thickness 0.25mm, as rolled, 99.99+%,
GF50091757 Cadmium, foil, 10mm disks, thickness 0.5mm, as rolled, 99.95%,
GF45457960 Cadmium, foil, 10mm disks, thickness 1.0mm, as rolled, 100%,
GF60281737 Cadmium, foil, 10mm disks, thickness 1.0mm, as rolled, 99.95%,
GF21075184 Cadmium, foil, 15mm disks, thickness 0.005mm, 99.7+%,
GF36899937 Cadmium, foil, 15mm disks, thickness 0.015mm, 99.7+%,
GF39261843 Cadmium, foil, 15mm disks, thickness 0.01mm, 99.7+%,
GF46599407 Cadmium, foil, 15mm disks, thickness 0.025mm, 99.7+%,
GF63236213 Cadmium, foil, 15mm disks, thickness 0.05mm, as rolled, 99.99+%,
GF83449097 Cadmium, foil, 15mm disks, thickness 0.125mm, as rolled, 99.95%,
GF29402377 Cadmium, foil, 15mm disks, thickness 0.125mm, as rolled, 99.99+%,
GF33887885 Cadmium, foil, 15mm disks, thickness 0.25mm, as rolled, 99.9%,
GF72666546 Cadmium, foil, 15mm disks, thickness 0.25mm, as rolled, 99.95%,
GF09346563 Cadmium, foil, 15mm disks, thickness 0.25mm, as rolled, 99.99+%,
GF27095205 Cadmium, foil, 15mm disks, thickness 0.5mm, as rolled, 99.95%,
GF82245708 Cadmium, foil, 15mm disks, thickness 1.0mm, as rolled, 100%,
GF98852260 Cadmium, foil, 15mm disks, thickness 1.0mm, as rolled, 99.95%,
GF69270824 Cadmium, foil, 1m coil, thickness 0.5mm, as rolled, 100%,
GF04527132 Cadmium, foil, 25mm disks, thickness 0.05mm, as rolled, 99.99+%,
GF31136431 Cadmium, foil, 25mm disks, thickness 0.125mm, as rolled, 99.95%,
GF75296469 Cadmium, foil, 25mm disks, thickness 0.125mm, as rolled, 99.99+%,
GF35985370 Cadmium, foil, 25mm disks, thickness 0.25mm, as rolled, 99.9%,
GF24841953 Cadmium, foil, 25mm disks, thickness 0.25mm, as rolled, 99.95%,
GF86979986 Cadmium, wire reel, 1m, diameter 0.5mm, hard, 99.95+%,
GF67082328 Cadmium, wire reel, 1m, diameter 0.5mm, hard, 99.99+%,
GF63053276 Cadmium, wire reel, 1m, diameter 0.70mm, as drawn, 99.99+%,
GF01474635 Cadmium, wire reel, 1m, diameter 1.2mm, as drawn, 99.99+%,
GF29720875 Cadmium, wire reel, 1m, diameter 0.125mm, as drawn, 99.9%,
GF79124441 Cadmium, wire reel, 1m, diameter 1.0mm, hard, 99.999%,
GF72498347 Cadmium, wire reel, 2m, diameter 0.5mm, hard, 99.95+%,
GF85290699 Cadmium, wire reel, 2m, diameter 0.5mm, hard, 99.99+%,
GF75801146 Cadmium, wire reel, 2m, diameter 0.70mm, as drawn, 99.99+%,
GF14719080 Cadmium, wire reel, 2m, diameter 1.2mm, as drawn, 99.99+%,
GF15109387 Cadmium, wire reel, 2m, diameter 0.125mm, as drawn, 99.9%,
GF49846056 Cadmium, wire reel, 5m, diameter 0.25mm, hard, 99.99+%,
GF62293585 Cadmium, wire reel, 5m, diameter 0.5mm, hard, 99.95+%,
GF61336997 Cadmium, wire reel, 5m, diameter 0.5mm, hard, 99.99+%,
GF88672786 Cadmium, wire reel, 5m, diameter 0.125mm, as drawn, 99.9%,
GF68320931 Cadmium, foil, 25mm disks, thickness 0.25mm, as rolled, 99.99+%,
GF22792394 Cadmium, foil, 25mm disks, thickness 0.5mm, as rolled, 99.95%,
GF80713706 Cadmium, foil, 25mm disks, thickness 1.0mm, as rolled, 100%,
GF95387866 Cadmium, foil, 25mm disks, thickness 1.0mm, as rolled, 99.95%,
GF58853638 Cadmium, foil, 2m coil, thickness 0.5mm, as rolled, 100%,
GF51515939 Cadmium, foil, 4mm disks, thickness 0.005mm, 99.7+%,
GF92345193 Cadmium, foil, 4mm disks, thickness 0.015mm, 99.7+%,
GF01578641 Cadmium, foil, 4mm disks, thickness 0.01mm, 99.7+%,
GF81565533 Cadmium, foil, 4mm disks, thickness 0.025mm, 99.7+%,
GF64685751 Cadmium, foil, 4mm disks, thickness 0.05mm, as rolled, 99.99+%,
GF04636100 Cadmium, foil, 4mm disks, thickness 0.125mm, as rolled, 99.95%,
GF58810507 Cadmium, foil, 4mm disks, thickness 0.125mm, as rolled, 99.99+%,
GF97910950 Cadmium, wire reel, 0.1m, diameter 0.125mm, as drawn, 99.9%,
GF14326466 Cadmium, foil, 4mm disks, thickness 0.25mm, as rolled, 99.9%,
GF10427756 Cadmium, foil, 4mm disks, thickness 0.25mm, as rolled, 99.95%,
GF42101525 Cadmium, foil, 4mm disks, thickness 0.25mm, as rolled, 99.99+%,
GF13705297 Cadmium, foil, 4mm disks, thickness 0.5mm, as rolled, 99.95%,
GF18286151 Cadmium, foil, 4mm disks, thickness 1.0mm, as rolled, 100%,
GF90978553 Cadmium, foil, 4mm disks, thickness 1.0mm, as rolled, 99.95%,
GF65892392 Cadmium, foil, 50mm disks, thickness 0.05mm, as rolled, 99.99+%,
GF92054684 Cadmium, foil, 50mm disks, thickness 0.125mm, as rolled, 99.95%,
GF12058062 Cadmium, foil, 50mm disks, thickness 0.125mm, as rolled, 99.99+%,
GF03549898 Cadmium, foil, 50mm disks, thickness 0.25mm, as rolled, 99.95%,
GF54411207 Cadmium, foil, 50mm disks, thickness 0.5mm, as rolled, 99.95%,
GF89212945 Cadmium, foil, 50mm disks, thickness 1.0mm, as rolled, 100%,
GF39491332 Cadmium, foil, 6mm disks, thickness 0.005mm, 99.7+%,
GF43148875 Cadmium, foil, 6mm disks, thickness 0.015mm, 99.7+%,
GF97330091 Cadmium, foil, 6mm disks, thickness 0.01mm, 99.7+%,
GF95692157 Cadmium, foil, 6mm disks, thickness 0.025mm, 99.7+%,
GF85719279 Cadmium, foil, 6mm disks, thickness 0.05mm, as rolled, 99.99+%,
GF79513544 Cadmium, foil, 6mm disks, thickness 0.125mm, as rolled, 99.95%,
GF51312653 Cadmium, foil, 6mm disks, thickness 0.125mm, as rolled, 99.99+%,
GF90807925 Cadmium, foil, 6mm disks, thickness 0.25mm, as rolled, 99.9%,
GF15424950 Cadmium, foil, 6mm disks, thickness 0.25mm, as rolled, 99.95%,
GF62690159 Cadmium, foil, 6mm disks, thickness 0.25mm, as rolled, 99.99+%,
GF53269757 Cadmium, foil, 6mm disks, thickness 0.5mm, as rolled, 99.95%,
GF85769308 Cadmium, foil, 6mm disks, thickness 1.0mm, as rolled, 100%,
GF22735492 Cadmium, foil, 6mm disks, thickness 1.0mm, as rolled, 99.95%,
GF36417348 Cadmium, foil, 8mm disks, thickness 0.005mm, 99.7+%,
GF93608769 Cadmium, foil, 8mm disks, thickness 0.015mm, 99.7+%,
GF66261719 Cadmium, foil, 8mm disks, thickness 0.01mm, 99.7+%,
GF61519787 Cadmium, foil, 8mm disks, thickness 0.025mm, 99.7+%,
GF14909643 Cadmium, foil, 8mm disks, thickness 0.05mm, as rolled, 99.99+%,
GF39192543 Cadmium, foil, 8mm disks, thickness 0.125mm, as rolled, 99.95%,
GF14081880 Cadmium, foil, 8mm disks, thickness 0.125mm, as rolled, 99.99+%,
GF50990968 Cadmium, foil, 8mm disks, thickness 0.25mm, as rolled, 99.9%,
GF31079066 Cadmium, foil, 8mm disks, thickness 0.25mm, as rolled, 99.95%,
GF36269398 Cadmium, foil, 8mm disks, thickness 0.25mm, as rolled, 99.99+%,
GF10057700 Cadmium, foil, 8mm disks, thickness 0.5mm, as rolled, 99.95%,
GF21163665 Cadmium, foil, 8mm disks, thickness 1.0mm, as rolled, 100%,
GF79164951 Cadmium, foil, 8mm disks, thickness 1.0mm, as rolled, 99.95%,
GF33725897 Cadmium, foil, light tested, 100x100mm, thickness 0.015mm, 99.7+%,
GF53798428 Cadmium, foil, light tested, 100x100mm, thickness 0.025mm, 99.7+%,
GF68517489 Cadmium, foil, light tested, 100x100mm, thickness 0.05mm, as rolled, 99.99+%,
GF76685754 Cadmium, foil, light tested, 25x25mm, thickness 0.005mm, 99.7+%,
GF03678082 Cadmium, foil, light tested, 25x25mm, thickness 0.015mm, 99.7+%,
GF88365640 Cadmium, foil, light tested, 25x25mm, thickness 0.01mm, 99.7+%,
GF04184322 Cadmium, foil, light tested, 25x25mm, thickness 0.025mm, 99.7+%,
GF15869986 Cadmium, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99.99+%,
GF98886223 Cadmium, foil, light tested, 50x50mm, thickness 0.015mm, 99.7+%,
GF27412309 Cadmium, foil, light tested, 50x50mm, thickness 0.01mm, 99.7+%,
GF48275905 Cadmium, foil, light tested, 50x50mm, thickness 0.025mm, 99.7+%,
GF83748134 Cadmium, foil, light tested, 50x50mm, thickness 0.05mm, as rolled, 99.99+%,
GF47753691 Cadmium, foil, not light tested, 100x100mm, thickness 0.005mm, 99.7+%,
GF13871527 Cadmium, foil, not light tested, 100x100mm, thickness 0.015mm, 99.7+%,
GF75237138 Cadmium, foil, not light tested, 100x100mm, thickness 0.01mm, 99.7+%,
GF38539929 Cadmium, foil, not light tested, 100x100mm, thickness 0.025mm, 99.7+%,
GF89292640 Cadmium, foil, not light tested, 25x25mm, thickness 0.001mm, 99.7+%,
GF09311543 Cadmium, foil, not light tested, 25x25mm, thickness 0.002mm, 99.7+%,
GF37906746 Cadmium, foil, not light tested, 25x25mm, thickness 0.005mm, 99.7+%,
GF97223250 Cadmium, foil, not light tested, 25x25mm, thickness 0.015mm, 99.7+%,
GF32202582 Cadmium, foil, not light tested, 25x25mm, thickness 0.025mm, 99.7+%,
GF02263799 Cadmium, foil, not light tested, 50x50mm, thickness 0.002mm, 99.7+%,
GF38760420 Cadmium, foil, not light tested, 50x50mm, thickness 0.015mm, 99.7+%,
GF86774428 Cadmium, foil, not light tested, 50x50mm, thickness 0.01mm, 99.7+%,
GF85239096 Cadmium, rod, 1000mm, diameter 5.0mm, as drawn, 99.99+%,
GF79955960 Cadmium, rod, 100mm, diameter 10.0mm, 99.999%,
GF49573884 Cadmium, rod, 100mm, diameter 40mm, 99.9%,
GF03378652 Cadmium, rod, 100mm, diameter 5.0mm, as drawn, 99.99+%,
GF53078182 Cadmium, rod, 150mm, diameter 10.0mm, as drawn, 99.99+%,
GF94495214 Cadmium, rod, 200mm, diameter 10.0mm, 99.999%,
GF24530217 Cadmium, rod, 200mm, diameter 5.0mm, as drawn, 99.99+%,
GF04536626 Cadmium, rod, 25mm, diameter 40mm, 99.9%,
GF56177589 Cadmium, rod, 300mm, diameter 10.0mm, as drawn, 99.99+%,
GF31881911 Cadmium, rod, 500mm, diameter 5.0mm, as drawn, 99.99+%,
GF79039648 Cadmium, rod, 50mm, diameter 10.0mm, 99.999%,
GF72955772 Cadmium, rod, 50mm, diameter 40mm, 99.9%,
GF66301983 Cadmium, tube, 1000mm, outside diameter 2.29mm, inside diameter 1.27mm, wall thickness 0.51mm, 99.95+%,
GF86338154 Cadmium, tube, 100mm, outside diameter 2.29mm, inside diameter 1.27mm, wall thickness 0.51mm, 99.95+%,
GF92269019 Cadmium, tube, 200mm, outside diameter 2.29mm, inside diameter 1.27mm, wall thickness 0.51mm, 99.95+%,
GF33627023 Cadmium, tube, 500mm, outside diameter 2.29mm, inside diameter 1.27mm, wall thickness 0.51mm, 99.95+%,
GF84097794 Cadmium, wire reel, 0.1m, diameter 0.25mm, hard, 99.99+%,
GF84184151 Cadmium, wire reel, 0.1m, diameter 0.5mm, hard, 99.95+%,
GF86697714 Cadmium, wire reel, 0.1m, diameter 0.5mm, hard, 99.99+%,
GF66689522 Cadmium, foil, not light tested, 50x50mm, thickness 0.005mm, 99.7+%,
GF83538355 Cadmium, foil, not light tested, 25x25mm, thickness 0.01mm, 99.7+%,
348600 Cadmium, wire, diam. 1.0 mm, 99.999% trace metals basis,