• USA Home
  • Search Results

  • cadmiumiodide36622779080911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cadmium iodide

4 Products
Synonym:   Cadmium diiodide
Image of Cadmium iodide
CAS Number:
Empirical Formula (Hill Notation):
 CdI2
Molecular Weight:
 366.22
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
  Solubility
Show More