• USA Home
  • Search Results

  • caffeicacid1801633139511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Caffeic acid

2 Products
Synonym:   3,4-Dihydroxybenzeneacrylic acid, 3,4-Dihydroxycinnamic acid, 3-(3,4-Dihydroxyphenyl)-2-propenoic acid
Image of Caffeic acid
CAS Number:
Linear Formula:
 (HO)2C6H3CH=CHCO2H
Molecular Weight:
 180.16
 Beilstein/REAXYS Number:
 1954563
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Suitability
  Color
  Special Grade
  Physical Form
Show More