• USA Home
  • Search Results

  • caffeine194195808211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Caffeine

14 Products
Synonym:   7-Methyltheobromine, 1,3,7-Trimethylxanthine, 3,7-Dihydro-1,3,7-trimethyl-1H-purine-2,6-dione
Image of Caffeine
CAS Number:
Linear Formula:
 C₈H₁₀N₄O₂
Molecular Weight:
 194.19 g/mol
 EC Number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Titertest
  Purity Grade
  Melting Point
  Product Line
  Anal. Standard Form
  Suitability
  Assay
  Solubility
  Special Grade
Show More