• USA Home
  • Search Results

  • caffeinemeltingpointstandard194195808211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Caffeine melting point standard

2 Products
Synonym:   1,3,7-Trimethylxanthine, Caffeine
Image of Caffeine melting point standard
CAS Number:
Empirical Formula (Hill Notation):
 C8H10N4O2
Molecular Weight:
 194.19
 Beilstein/REAXYS Number:
 17705
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Physical Form
Show More