• USA Home
  • Search Results

  • caffeinesolution123455808211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Caffeine solution

2 Products
Image of Caffeine solution
CAS Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Datasheet (PDF)  |  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Anal. Standard Form
  Special Grade
  Titertest
Show More