• USA Home
  • Search Results

  • caftaricacid312236787958711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Caftaric acid

4 Products
Synonym:   2-Caffeoyl-L-tartaric acid
Image of Caftaric acid
CAS Number:
Empirical Formula (Hill Notation):
 C13H12O9
Molecular Weight:
 312.23
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Titertest
Show More