• USA Home
  • Search Results

  • calcitoninsalmon3431854793185111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Calcitonin salmon

4 Products
Synonym:   Thyrocalcitonin
Image of Calcitonin salmon
CAS Number:
Empirical Formula (Hill Notation):
 C145H240N44O48S2
Molecular Weight:
 3431.85
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Manufacturer designation
  Special Grade
  Solubility
  Ice Shipping
  Assay
  Physical Form
Show More