• USA Home
  • Search Results

  • calciumacetate158176254411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Calcium acetate

2 Products
Image of Calcium acetate
CAS Number:
Linear Formula:
 (CH3CO2)2Ca
Molecular Weight:
 158.17
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Application(s)
Show More