• USA Home
  • Search Results

  • calciumcarbide64107520711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Calcium carbide

2 Products
Image of Calcium carbide
CAS Number:
Linear Formula:
 CaC2
Molecular Weight:
 64.10
 Beilstein/REAXYS Number:
 3909011
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Special Grade
  Assay
Show More