• USA Home
  • Search Results

  • calciumchloride110981004352411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Calcium chloride

15 Products
Image of Calcium chloride
CAS Number:
Linear Formula:
 CaCl2
Molecular Weight:
 110.98
 EC Number:
 MDL number:
 NACRES:
 NA.55
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Vapor pressure
  Application(s)
  Restriction
  Solubility
  Quality clarifier
  pH Value
  Potency
  Particle Dimension
  Melting Point
  Density
  Product Line
  Special Grade
  Physical Form
Show More

Product #
Description
C1016 Calcium chloride, anhydrous, granular, ≤7.0 mm, ≥93.0%,
C4901 Calcium chloride, anhydrous, powder, ≥97%,
C5670 Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%,
499609 Calcium chloride, anhydrous, powder, 99.99% trace metals basis,
449709 Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis,
1.02378 Calcium chloride, anhydrous granules Reag. Ph Eur,
746495 Calcium chloride, anhydrous, free-flowing, Redi-Dri, ≥97%,
793639 Calcium chloride, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥96%,
1.02391 Calcium chloride, anhydrous, granular ~ 2 - 6 mm,
383147 Calcium chloride, desiccant, ACS reagent, ≥96.0%,
1.02379 Calcium chloride, anhydrous, granular ~ 1-2 mm,
902179 Calcium chloride, anhydrous, granular, ≤7.0 mm, free-flowing, Redi-Dri, ReagentPlus®, ≥93%,
429759 Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis,
1.02392 Calcium chloride, anhydrous, granular ~ 6 - 14 mm,
1.02083 Calcium chloride, fused, granular about 0.5-2.0 mm for elementary analysis,