• USA Home
  • Search Results

  • calciumchloride147011003504811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Calcium chloride

2 Products
Synonym:   Calcium chloride dihydrate
Image of Calcium chloride
CAS Number:
Empirical Formula (Hill Notation):
 CaCl2 · 2H2O
Molecular Weight:
 147.01
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More