• USA Home
  • Search Results

  • calciumchloridedihydrate147011003504811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Calcium chloride dihydrate

17 Products
Synonym:   Calcium dichloride
Image of Calcium chloride dihydrate
CAS Number:
Empirical Formula (Hill Notation):
 CaCl2 · 2H2O
Molecular Weight:
 147.01
 E Number:
 E509
 EC Index Number:
 233-140-8
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  UV-Absorption Condition
  Purity Grade
  UV-Absorption Values
  Product Line
  Application(s)
  Potency
  pH Value
  Solubility
  Special Grade
  Agency/Method
Show More

Product #
Description
223506 Calcium chloride dihydrate, ACS reagent, ≥99%,
C3306 Calcium chloride dihydrate, for molecular biology, ≥99.0%,
C7902 Calcium chloride dihydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%,
C3881 Calcium chloride dihydrate, ReagentPlus®, ≥99.0%,
C5080 Calcium chloride dihydrate, BioXtra, ≥99.0%,
31307 Calcium chloride dihydrate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%,
21097 Calcium chloride dihydrate, BioUltra, for molecular biology, ≥99.5% (KT),
12022 Calcium chloride dihydrate, puriss., meets analytical specification of Ph. Eur., USP, FCC, E509, 99-103%, ≤0.0001% Al,
21098 Calcium chloride dihydrate, BioUltra, ≥99.5% (KT),
21102 Calcium chloride dihydrate, tested according to Ph Eur,
1.72580 Calcium chloride dihydrate, extra pure, for table water FCC,E 509,
1.42002 Calcium chloride dihydrate, EMPROVE® ESSENTIAL Ph Eur,BP,JP,USP,FCC,E 509,
1.01867 Calcium chloride dihydrate, cryst., Ph Eur,JP,USP,FCC,E 509,
1.02465 Calcium chloride dihydrate, EMPROVE® API Ph Eur,BP,JP,USP,
1.37101 Calcium chloride dihydrate, EMPROVE® EXPERT Ph Eur,BP,JP,USP,
1.02382 Calcium chloride dihydrate, for analysis EMSURE® ACS,Reag. Ph Eur,
208290 Calcium chloride dihydrate, ACS Reagent Grade.,