• USA Home
  • Search Results

  • calciumdgluconate4303729928511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Calcium D-gluconate

1 Products
Synonym:   2,3,4,5,6-Pentahydroxycaproic Acid hemicalcium salt, Gluconic acid hemicalcium salt
Image of Calcium <SC>D</SC>-gluconate
CAS Number:
Empirical Formula (Hill Notation):
 C12H22CaO14
Molecular Weight:
 430.37
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More