• USA Home
  • Search Results

  • calciumgluconateanhydrous4303729928511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Calcium gluconate anhydrous

2 Products
Synonym:   2,3,4,5,6-Pentahydroxycaproic Acid hemicalcium salt, Calcium D-gluconate, Gluconic acid hemicalcium salt
Image of Calcium gluconate anhydrous
CAS Number:
Empirical Formula (Hill Notation):
 C12H22CaO14
Molecular Weight:
 430.37
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
Show More