• USA Home
  • Search Results

  • calciumiodide293891010268811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Calcium iodide

3 Products
Synonym:   Calcium diiodide
Image of Calcium iodide
CAS Number:
Linear Formula:
 CaI2
Molecular Weight:
 293.89
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Particle Dimension
  Physical Form
Show More