• USA Home
  • Search Results

  • calciumnitratetetrahydrate236151347734411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Calcium nitrate tetrahydrate

4 Products
Image of Calcium nitrate tetrahydrate
CAS Number:
Linear Formula:
 Ca(NO3)2 · 4H2O
Molecular Weight:
 236.15
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  Suitability
  Purity Grade
  pH Value
  Special Grade
Show More