• USA Home
  • Search Results

  • calciumoxide5608130578811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Calcium oxide

5 Products
Synonym:   Lime, Quicklime
Image of Calcium oxide
CAS Number:
Linear Formula:
 CaO
Molecular Weight:
 56.08
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Purity Grade
  Restriction
  Loss
  Physical Form
  Special Grade
Show More