• USA Home
  • Search Results

  • calciumphosphatemonobasicmonohydrate252071003130811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Calcium phosphate monobasic monohydrate

1 Products
Synonym:   Calcium bis(dihydrogenphosphate) monohydrate, Calcium dihydrogenphosphate monohydrate, Primary calcium phosphate monohydrate
Image of Calcium phosphate monobasic monohydrate
CAS Number:
Linear Formula:
 Ca(H2PO4)2 · H2O
Molecular Weight:
 252.07
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
Show More