• USA Home
  • Search Results

  • calciumsulfatedihydrate172171010141411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Calcium sulfate dihydrate

5 Products
Image of Calcium sulfate dihydrate
CAS Number:
Linear Formula:
 CaSO4 · 2H2O
Molecular Weight:
 172.17
 EC Number:
 MDL number:
 NACRES:
 NA.55
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Reaction Suitability
  Agency/Method
  pH Value
  Solubility
  Potency
  Density
  Bulk Density
  Purity Grade
  Special Grade
  Product Line
  Physical Form
Show More