• USA Home
  • Search Results

  • calmodulinfrombovinetestes123457710746111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Calmodulin from bovine testes

1 Products
Synonym:   CaM, Phosphodiesterase 3′:5′-cyclic nucleotide activator
Image of Calmodulin from bovine testes
CAS Number:
 MDL number:
 NACRES:
 NA.61
Popular Documents:  Datasheet (PDF)  |  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More