• USA Home
  • Search Results

  • calycosin284262057557911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Calycosin

3 Products
Synonym:   3′,7-Dihydroxy-4′-methoxy-isoflavone, 3′-Hydroxyformononetin, Cyclosin
Image of Calycosin
CAS Number:
Empirical Formula (Hill Notation):
 C16H12O5
Molecular Weight:
 284.26
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Manufacturer designation
Show More