• USA Home
  • Search Results

  • camphor152237622211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(±)-Camphor

4 Products
Synonym:   1,7,7-Trimethylbicyclo[2.2.1]heptan-2-one
Image of (±)-Camphor
CAS Number:
Empirical Formula (Hill Notation):
 C10H16O
Molecular Weight:
 152.23
 Beilstein/REAXYS Number:
 1907611
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Suitability
  Special Grade
  Quality clarifier
  Assay
Show More